(Propargyloxy)trimethylsilane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trimethyl(prop-2-yn-1-yloxy)silane
L12936-06
L12936-14
CAAAAL12936-06EA
69.68
CAD
CAAAAL12936-06
CAAAAL12936-14
(Propargyloxy)trimethylsilane 97%
(Propargyloxy)trimethylsilane
Purity: 97%
CAS No.: 5582-62-7
Molecular Formula: C6H12OSi
Formula Weight: 128.25
Storage Temperature: 2°C to 8°C
Flash Point: 8°(46°F)
Refractive Index: 1.4080
Boiling Point: 110-111°
Beilstein Reference: 2036793
UN No.: 1993
MDL No.: MFCD00042844
Formula:
C₆H₁₂OSi Boiling Pt: 110…111 °C Density: 0.833 Flash Pt: 8 °C(46 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00042844 CAS Number: 5582-62-7 EINECS: 226-981-7 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...