Domiphen bromide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (Dodecyldimethyl-2-phenoxyethyl)ammonium bromide, N,N-Dimethyl-N-(2-phenoxyethyl)dodecan-1-aminium bromide

L12930-14 L12930-22
CAAAAL12930-14EA 82.16 CAD
CAAAAL12930-14 CAAAAL12930-22
Domiphen bromide 97%
Domiphen bromide

Purity: 97% CAS No.: 538-71-6 Molecular Formula: C22H40BrNO Formula Weight: 414.48 Melting Point: 117-118° Beilstein Reference: 3922742 MDL No.: MFCD00011769

Formula: C₂₂H₄₀BrNO
MW: 414.48 g/mol
Melting Pt: 117…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00011769
CAS Number: 538-71-6
EINECS: 208-702-0
Merck Index: 14,3415

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR