2,3-Dimethylmaleic anhydride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12919-06
L12919-14
CAAAAL12919-06EA
133.35
CAD
CAAAAL12919-06
CAAAAL12919-14
2,3-Dimethylmaleic anhydride 97%
2,3-Dimethylmaleic anhydride
Purity: 97%
CAS No.: 766-39-2
Molecular Formula: C6H6O3
Formula Weight: 126.11
Melting Point: 93-96°
Boiling Point: 222-223°
MDL No.: MFCD00005523
Formula:
C₆H₆O₃ Boiling Pt: 222…223 °C Melting Pt: 93…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005523 CAS Number: 766-39-2 EINECS: 212-165-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...