2-Hydroxy-4-methoxy-5-sulfobenzophenone hydrate tech. 85% may cont. up to 10% 2-propanol, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-4-methoxybenzophenone-5-sulfonic Acid Hydrate, 2-Hydroxy-4-methoxybenzophenone-5-sulphonic Acid Hydrate

L12912-09 L12912-18
CAAAAL12912-09EA 117.71 CAD
CAAAAL12912-09 CAAAAL12912-18
2-Hydroxy-4-methoxy-5-sulfobenzophenone hydrate tech. 85% may cont. up to 10% 2-propanol, Technical Grade
2-Hydroxy-4-methoxy-5-sulfobenzophenone hydrate

Purity: 85% CAS No.: 4065-45-6 Molecular Formula: C14H12O6S.xH2O Formula Weight: 308.31(anhy) Melting Point: 108-112° Beilstein Reference: 2889165 Merck Reference: 14,8983 MDL No.: MFCD00977409

Formula: C₁₄H₁₂O₆S·H₂O
MW: 308.31 g/mol (anhy)
Melting Pt: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00024962
CAS Number: 4065-45-6
EINECS: 223-772-2
Merck Index: 14,8983

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR