2-(Benzyloxy)benzaldehyde 98%
Supplier: Thermo Scientific Chemicals
L12866-09
L12866-18
CAAAAL12866-09EA
60.67
CAD
CAAAAL12866-09
CAAAAL12866-18
2-(Benzyloxy)benzaldehyde 98%
2-(Benzyloxy)benzaldehyde
Purity: 98%
CAS No.: 5896-17-3
Molecular Formula: C14H12O2
Formula Weight: 212.25
Melting Point: 46-47°
Beilstein Reference: 780307
Formula:
C₁₄H₁₂O₂ Melting Pt: 46…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016583 CAS Number: 5896-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...