3-(Methylthio)propionic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-(Methylthio)propionic Acid
L12861-09
L12861-18
CAAAAL12861-09EA
59.41
CAD
CAAAAL12861-09
CAAAAL12861-18
3-(Methylthio)propionic acid 98%
3-(Methylthio)propionic acid
Purity: 98%
CAS No.: 646-01-5
Molecular Formula: C4H8O2S
Formula Weight: 120.17
Flash Point: 104°(219°F)
Refractive Index: 1.4885
Odor: Unpleasant
Melting Point: ca 16°
Boiling Point: 244-247°
Beilstein Reference: 1743054
UN No.: 3265
MDL No.: MFCD00059635
Formula:
C₄H₈O₂S Boiling Pt: 244…247 °C Melting Pt: ca 16 °C Density: 1.16 Flash Pt: 104 °C(219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00059635 CAS Number: 646-01-5 EINECS: 211-460-9 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...