Hydroxypivalic acid ≥97%
Supplier: Thermo Scientific Chemicals
L12858-18
L12858-30
CAAAAL12858-18EA
107.76
CAD
CAAAAL12858-18
CAAAAL12858-30
Hydroxypivalic acid ≥97%
Hydroxypivalic acid
Purity: 98%
CAS No.: 4835-90-9
Molecular Formula: C5H10O3
Formula Weight: 118.13
Odor: Odorless
Melting Point: 122-126°
Beilstein Reference: 1747025
MDL No.: MFCD00059953
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...