Divinyl sulfone 97% stabilised

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with hydroquinone (HQ) 0.05%

L12827-09 L12827-18
CAAAAL12827-09EA 107.91 CAD
CAAAAL12827-09 CAAAAL12827-18
Divinyl sulfone 97% stabilised
Divinyl sulphone

Fieser: 7,23


Purity: 97% CAS No.: 77-77-0 Molecular Formula: C4H6O2S Formula Weight: 118.16 Storage Temperature: 2°C to 8°C Flash Point: 102°(215°F) Refractive Index: 1.4760 Melting Point: ca -26° Boiling Point: 233-234° Beilstein Reference: 1071329 UN No.: 2927 MDL No.: MFCD00008623

Formula: C₄H₆O₂S
Boiling Pt: 233…234 °C
Melting Pt: ca …26 °C
Density: 1.177
Flash Pt: 102 °C(215 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008623
CAS Number: 77-77-0
EINECS: 201-057-6
UN: 2927
ADR: "6.1",II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR