3-(4-Morpholino)propyl isothiocyanate 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12819-03
L12819-06
CAAAAL12819-03EA
44.71
CAD
CAAAAL12819-03
CAAAAL12819-06
3-(4-Morpholino)propyl isothiocyanate 96%
3-(4-Morpholino)propyl isothiocyanate
Purity: 96%
CAS No.: 32813-50-6
Molecular Formula: C8H14N2OS
Formula Weight: 186.28
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5305
Boiling Point: 182-183°/13mm
Beilstein Reference: 879217
UN No.: 2922
MDL No.: MFCD00041222
Formula:
C₈H₁₄N₂OS Boiling Pt: 182…183 °C/13 mm Storage Temperature: Refrigerator |
MDL Number:
MFCD00041222 CAS Number: 32813-50-6 UN: 2922 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...