2-Amino-4-hydroxyquinoline hydrate, (max. 10% H₂O) 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12760-03
L12760-06
CAAAAL12760-03EA
44.24
CAD
CAAAAL12760-03
CAAAAL12760-06
2-Amino-4-hydroxyquinoline hydrate, (max. 10% H₂O) 97%
2-Amino-4-hydroxyquinoline hydrate
Purity: 97%
CAS No.: 42712-64-1
Molecular Formula: C9H8N2O.xH2O
Formula Weight: 160.18(anhy)
Melting Point: ca 298° dec.
Beilstein Reference: 124785
MDL No.: MFCD00052386
Formula:
C₉H₈N₂O·xH₂O Melting Pt: ∼ 298 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00052386 CAS Number: 42712-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...