Isobutyl acrylate 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Acrylic Acid Isobutyl Ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 100 ppm
L12739-AE
L12739-AP
CAAAAL12739-AEEA
66.2
CAD
CAAAAL12739-AE
CAAAAL12739-AP
Isobutyl acrylate 99% stabilized
Isobutyl acrylate
stab with 100ppm 4-methoxyphenol
Purity: 99%
CAS No.: 106-63-8
Molecular Formula: C7H12O2
Formula Weight: 128.17
Flash Point: 32°(89°F)
Refractive Index: 1.4150
Melting Point: -61°
Boiling Point: 139°
Beilstein Reference: 1749388
UN No.: 2527
MDL No.: MFCD00042865
Formula:
C₇H₁₂O₂ MW: 128.17 g/mol Boiling Pt: 139 °C Melting Pt: –61 °C Density: 0.888 Flash Pt: 32 °C(89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042865 CAS Number: 106-63-8 EINECS: 203-417-8 UN: 2527 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...