4-Bromo-2-ethylaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12725-06
L12725-14
CAAAAL12725-06EA
76.63
CAD
CAAAAL12725-06
CAAAAL12725-14
4-Bromo-2-ethylaniline 97%
4-Bromo-2-ethylaniline
Purity: 97%
CAS No.: 45762-41-2
Molecular Formula: C8H10BrN
Formula Weight: 200.08
Refractive Index: 1.6020
Boiling Point: 110-112°/2mm
UN No.: 2810
MDL No.: MFCD00041434
Formula:
C₈H₁₀BrN Boiling Pt: 110…112 °C/2 mm Density: 1.412 Storage Temperature: Ambient |
MDL Number:
MFCD00041434 CAS Number: 45762-41-2 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...