Diacetin mixture of isomers ∼50% remainder triacetin and monoacetin, tech.
Supplier: Thermo Scientific Chemicals
L12708-0B
L12708-30
CAAAAL12708-0BEA
146.47
CAD
CAAAAL12708-0B
CAAAAL12708-30
Diacetin mixture of isomers ∼50% remainder triacetin and monoacetin, tech.
Diacetin
CAS No.: 25395-31-7
Molecular Formula: C7H12O5
Formula Weight: 176.17
Flash Point: 141°(285°F)
Refractive Index: 1.4440
Boiling Point: 250-280° dec.
Beilstein Reference: 1706903
MDL No.: MFCD00008717
Formula:
C₇H₁₂O₅ Boiling Pt: 250…280 °C (dec.) Density: 1.190 Flash Pt: 141 °C(285 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008717 CAS Number: 25395-31-7 EINECS: 246-941-2 Merck Index: 14,2961 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...