TBPE (=tetrabromophenolphthalein ethyl ester potassium salt) ≥97%

Supplier: Thermo Scientific Chemicals

Synonyms: 3',3'',5',5''-Tetrabromophenolphthalein ethyl ester potassium salt, Tetrabromophenolphthalein ethyl ester potassium salt, Tetrabromophenolphthalein Ethyl Ester Potassium Salt

L12674-03 L12674-06
CAAAAL12674-03EA 106.33 CAD
CAAAAL12674-03 CAAAAL12674-06
TBPE (=tetrabromophenolphthalein ethyl ester potassium salt) ≥97%
TBPE (=tetrabromophenolphthalein ethyl ester potassium salt)

CAS No.: 62637-91-6 Molecular Formula: C22H13Br4KO4 Formula Weight: 700.06 Melting Point: ca 273° dec. Beilstein Reference: 3848263 MDL No.: MFCD00011662

Formula: C₂₂H₁₄Br₄O₄
Melting Pt: ca 273 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011662
CAS Number: 62637-91-6
EINECS: 263-661-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR