4-Bromo-2-chloro-6-methylaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

L12590-06 L12590-14
CAAAAL12590-06EA 93.38 CAD
CAAAAL12590-06 CAAAAL12590-14
4-Bromo-2-chloro-6-methylaniline 98%
4-Bromo-2-chloro-6-methylaniline

Purity: 98% CAS No.: 30273-42-8 Molecular Formula: C7H7BrClN Formula Weight: 220.50 Melting Point: 37-40° Beilstein Reference: 2717133 UN No.: 2811 MDL No.: MFCD00041432

Formula: C₇H₇BrClN
MW: 220.50 g/mol
Melting Pt: 37…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00041432
CAS Number: 30273-42-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR