2-(2,2,2-Trifluoroacetyl)thiophene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Trifluoroacetyl)thiophene

L12585-03 L12585-06
CAAAAL12585-03EA 56.41 CAD
CAAAAL12585-03 CAAAAL12585-06
2-(2,2,2-Trifluoroacetyl)thiophene 98%
2-(2,2,2-Trifluoroacetyl)thiophene

Purity: 98% CAS No.: 651-70-7 Molecular Formula: C6H3F3OS Formula Weight: 180.15 Flash Point: 58°(136°F) Refractive Index: 1.4845 Boiling Point: 163-165° Beilstein Reference: 1309208 UN No.: 1993 MDL No.: MFCD00041427

Formula: C₆H₅ClOS
MW: 180.15 g/mol
Boiling Pt: 163…165 °C
Density: 1.403
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041427
CAS Number: 651-70-7
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR