N,N'-Ethylenebisacrylamide 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N,N-(Ethane-1,2-diyl)diacrylamide
L12544-03
L12544-06
CAAAAL12544-03EA
119.13
CAD
CAAAAL12544-03
CAAAAL12544-06
N,N'-Ethylenebisacrylamide 96%
N,N'-Ethylenebisacrylamide
Purity: 96%
CAS No.: 2956-58-3
Molecular Formula: C8H12N2O2
Formula Weight: 168.20
Storage Temperature: 2°C to 8°C
Melting Point: 141-143°
Beilstein Reference: 1771570
MDL No.: MFCD00042935
Formula:
C₈H₁₂N₂O₂ MW: 168.20 g/mol Melting Pt: 141…143 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00042935 CAS Number: 2956-58-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...