Ethyl L-2-isothiocyanatopropionate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12532-03
L12532-06
CAAAAL12532-03EA
116.45
CAD
CAAAAL12532-03
CAAAAL12532-06
Ethyl L-2-isothiocyanatopropionate 97%
Ethyl L-2-isothiocyanatopropionate
Purity: 97%
CAS No.: 39574-16-8
Molecular Formula: C6H9NO2S
Formula Weight: 159.20
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4925
Boiling Point: 63-64°/1mm
Beilstein Reference: 1723243
UN No.: 2922
MDL No.: MFCD00070636
Formula:
C₆H₉NO₂S Boiling Pt: 63…64 °C/1 mm Storage Temperature: Refrigerator |
MDL Number:
MFCD09025691 CAS Number: 39574-16-8 UN: 2922 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...