N,N-Bis(trifluoromethanesulfonyl)aniline 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12531-06
L12531-14
CAAAAL12531-03EA
44.21
CAD
CAAAAL12531-03
CAAAAL12531-06
CAAAAL12531-14
N,N-Bis(trifluoromethanesulfonyl)aniline 99%
N,N-Bis(trifluoromethanesulphonyl)aniline
Fieser: 12,395 13,248 18,292
Purity: 99%
CAS No.: 37595-74-7
Molecular Formula: (CF3SO2)2NC6H5
Formula Weight: 357.25
Melting Point: 100-102°
Beilstein Reference: 1269141
Formula:
C₈H₅F₆NO₄S₂ Melting Pt: 100…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000404 CAS Number: 37595-74-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...