Ethyl-4-isothiocyanatobenzoate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-(Ethoxycarbonyl)phenyl isothiocyanate
L12522-04
L12522-09
CAAAAL12522-04EA
61.3
CAD
CAAAAL12522-04
CAAAAL12522-09
Ethyl-4-isothiocyanatobenzoate 97%
Ethyl 4-isothiocyanatobenzoate
Purity: 97%
CAS No.: 1205-06-7
Molecular Formula: C10H9NO2S
Formula Weight: 207.25
Odor: Irritating
Melting Point: 52-56°
Boiling Point: 123-127°/1mm
Beilstein Reference: 2213400
UN No.: 2923
MDL No.: MFCD00022058
Formula:
C₁₀H₉NO₂S MW: 207.25 Boiling Pt: 123…127 °C/1 mm Melting Pt: 52…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022058 CAS Number: 1205-06-7 EINECS: 214-880-0 UN: 2923 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...