2-Bromoethyl acrylate 94% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 900 - 1500 ppm
L12502-03
L12502-06
CAAAAL12502-03EA
86.9
CAD
CAAAAL12502-03
CAAAAL12502-06
2-Bromoethyl acrylate 94% stabilized
2-Bromoethyl acrylate
Stab. with 0.1% 4-methoxyphenol
Purity: 94%
CAS No.: 4823-47-6
Molecular Formula: CH2=CHCO2CH2CH2Br
Formula Weight: 179.02
Storage Temperature: 2°C to 8°C
Boiling Point: 52-53°/5mm
Beilstein Reference: 1749713
MDL No.: MFCD00013541
Formula:
CH₂=CHCO₂CH₂CH₂Br MW: 179.02 g/mol Boiling Pt: 52…53 °C/5 mm Storage Temperature: Refrigerator |
MDL Number:
MFCD00013541 CAS Number: 4823-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...