Sodium hydrogen-L-tartrate, anhydrous 98%
Supplier: Thermo Scientific Chemicals
L12469-14
L12469-22
L12469-36
CAAAAL12469-14EA
47.56
CAD
CAAAAL12469-14
CAAAAL12469-22
CAAAAL12469-36
Sodium hydrogen-L-tartrate, anhydrous 98%
Sodium hydrogen-L-tartrate
Purity: 98%
CAS No.: 526-94-3
Molecular Formula: C4H5NaO6
Formula Weight: 172.07
Melting Point: ca 243° dec.
Beilstein Reference: 6099125
Optical Rotation: +24.5° (c=3 in water)
MDL No.: MFCD00065393
Formula:
C₄H₅NaO₆ Melting Pt: ca 243 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00065393 CAS Number: 526-94-3 EINECS: 208-400-9 Merck Index: 14,8589 |
Specification Test Results
Optical rotation | +24.5° (c=3 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...