2-Cyanophenylacetonitrile tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12419-06
L12419-14
CAAAAL12419-06EA
38.24
CAD
CAAAAL12419-06
CAAAAL12419-14
2-Cyanophenylacetonitrile tech. 90%, Technical Grade
2-Cyanophenylacetonitrile
Purity: 90%
CAS No.: 3759-28-2
Molecular Formula: C9H6N2
Formula Weight: 142.16
Melting Point: 73-79°
Beilstein Reference: 1940497
UN No.: 3439
MDL No.: MFCD00001895
Formula:
C₉H₆N₂ Melting Pt: 73…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001895 CAS Number: 3759-28-2 EINECS: 223-167-3 UN: 3439 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...