N-Benzoylthiourea 98%

Supplier: Thermo Scientific Chemicals
Warning

L12377-06 L12377-14
CAAAAL12377-06EA 46.45 CAD
CAAAAL12377-06 CAAAAL12377-14
N-Benzoylthiourea 98%
N-Benzoylthiourea

Purity: 98% CAS No.: 614-23-3 Molecular Formula: C8H8N2OS Formula Weight: 180.23 Odor: Odorless Melting Point: 174-176° Beilstein Reference: 1942772 MDL No.: MFCD00041191

Formula: C₈H₈N₂OS
Melting Pt: 174…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00041191
CAS Number: 614-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR