4-tert-Butylstyrene 94% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) 50 ppm
L12356-AE
L12356-AP
CAAAAL12356-AEEA
45.5
CAD
CAAAAL12356-AE
CAAAAL12356-AP
4-tert-Butylstyrene 94% stabilized
4-tert-Butylstyrene
Stab. with 50 ppm 4-tert-butylcatechol
Purity: 94%
CAS No.: 1746-23-2
Molecular Formula: C12H16
Formula Weight: 160.26
Flash Point: 80°(176°F)
Refractive Index: 1.5250
Melting Point: -38°
Boiling Point: 219°
Beilstein Reference: 1903714
MDL No.: MFCD00065126
Formula:
C₁₂H₁₆ Boiling Pt: 219 °C Melting Pt: –38 °C Density: 0.884 Flash Pt: 80 °C(176 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00065126 CAS Number: 1746-23-2 EINECS: 217-126-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...