1-Dimethylamino-2-propyne 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N,N-Dimethylpropargylamine, N,N-Dimethyl-2-propyn-1-amine
L12336-06
L12336-14
CAAAAL12336-06EA
56.25
CAD
CAAAAL12336-06
CAAAAL12336-14
1-Dimethylamino-2-propyne 98%
1-Dimethylamino-2-propyne
Purity: 98%
CAS No.: 7223-38-3
Molecular Formula: HC≡CCH2N(CH3)2
Formula Weight: 83.13
Flash Point: -7°(19°F)
Refractive Index: 1.4200
Boiling Point: 80-81°
Beilstein Reference: 1071222
UN No.: 2733
MDL No.: MFCD00008575
Formula:
C₅H₉N MW: 83.13 g/mol Boiling Pt: 80…81 °C Density: 0.772 Flash Pt: –7 °C (19 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008575 CAS Number: 7223-38-3 EINECS: 230-620-9 UN: 2733 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...