2,4-Diamino-6-nitroquinazoline 98%

Supplier: Thermo Scientific Chemicals
Warning

L12330-03 L12330-06
CAAAAL12330-03EA 56.09 CAD
CAAAAL12330-03 CAAAAL12330-06
2,4-Diamino-6-nitroquinazoline 98%
2,4-Diamino-6-nitroquinazoline

Purity: 98% CAS No.: 7154-34-9 Molecular Formula: C8H7N5O2 Formula Weight: 205.18 Melting Point: 360° Beilstein Reference: 749531

Formula: C₈H₇N₅O₂
Melting Pt: >360 °C
Storage Temperature: Ambient
MDL Number: MFCD00023910
CAS Number: 7154-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR