Cyclopropyl phenyl sulfide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12321-03
L12321-06
CAAAAL12321-03EA
43.77
CAD
CAAAAL12321-03
CAAAAL12321-06
Cyclopropyl phenyl sulfide 98+%
Cyclopropyl phenyl sulphide
Fieser: 5,372 6,319 7,190 8,138 9,271 10,138 11,231
Purity: 98+%
CAS No.: 14633-54-6
Molecular Formula: C9H10S
Formula Weight: 150.24
Flash Point: 91°(195°F)
Refractive Index: 1.5810
Odor: Stench
Boiling Point: 61-62°/1mm
Beilstein Reference: 2075701
UN No.: 3334
MDL No.: MFCD00009684
Formula:
C₉H₁₀S Boiling Pt: 61…62 °C/1 mm Density: 1.045 Flash Pt: 91 °C(195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009684 CAS Number: 14633-54-6 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...