Dodecylamine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dodecan-1-amine hydrochloride, Laurylamine Hydrochloride

L12314-09 L12314-18
CAAAAL12314-09EA 73.79 CAD
CAAAAL12314-09 CAAAAL12314-18
Dodecylamine hydrochloride 97%
Dodecylamine hydrochloride

Purity: 97% CAS No.: 929-73-7 Molecular Formula: C12H27N.HCl Formula Weight: 221.82 Melting Point: 186-188° Beilstein Reference: 3553943 MDL No.: MFCD00042017

Formula: C₁₂H₂₇N
Melting Pt: 186…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00042017
CAS Number: 929-73-7
EINECS: 213-205-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR