2-Nitro-5-(trifluoromethyl)aniline 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Amino-4-nitrobenzotrifluoride

L12310-03 L12310-06
CAAAAL12310-03EA 130.03 CAD
CAAAAL12310-03 CAAAAL12310-06
2-Nitro-5-(trifluoromethyl)aniline 97%
2-Nitro-5-(trifluoromethyl)aniline

Purity: 97% CAS No.: 402-14-2 Molecular Formula: C7H5F3N2O2 Formula Weight: 206.13 Melting Point: 98-102° Beilstein Reference: 2732344 UN No.: 2811 MDL No.: MFCD00042447

Formula: C₇H₅F₃N₂O₂
MW: 206.13 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00042447
CAS Number: 402-14-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR