β,3-Dinitrostyrene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12304-03
L12304-06
CAAAAL12304-03EA
116.76
CAD
CAAAAL12304-03
CAAAAL12304-06
β,3-Dinitrostyrene 98%
β,3-Dinitrostyrene
Purity: 98%
CAS No.: 882-26-8
Molecular Formula: C8H6N2O4
Formula Weight: 194.15
Melting Point: 125-127°
Beilstein Reference: 2051290
Formula:
C₈H₆N₂O₄ Melting Pt: 125…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052063 CAS Number: 882-26-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...