2-Hexyldecanoic acid 97%
Supplier: Thermo Scientific Chemicals
L12238-14
L12238-22
CAAAAL12238-14EA
47.56
CAD
CAAAAL12238-14
CAAAAL12238-22
2-Hexyldecanoic acid 97%
2-Hexyldecanoic acid
Purity: 97%
CAS No.: 25354-97-6
Molecular Formula: C16H32O2
Formula Weight: 256.43
Flash Point: 174°(345°F)
Refractive Index: 1.4460
Melting Point: 10°
Boiling Point: 165-168°/2mm
Beilstein Reference: 1777217
MDL No.: MFCD00059651
Formula:
C₁₆H₃₂O₂ Boiling Pt: 165…168 °C/2 mm Melting Pt: 10 °C Density: 0.880 Flash Pt: 174 °C(345 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00059651 CAS Number: 25354-97-6 EINECS: 246-885-9 Merck Index: 14,4709 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...