3-Trifluoromethylbutyric acid 97%

Supplier: Thermo Scientific Chemicals
Danger

L12160-03 L12160-06
CAAAAL12160-03EA 167.48 CAD
CAAAAL12160-03 CAAAAL12160-06
3-Trifluoromethylbutyric acid 97%
3-Trifluoromethylbutyric acid

Purity: 97% CAS No.: 348-75-4 Molecular Formula: C5H7F3O2 Formula Weight: 156.11 Refractive Index: 1.3590 Odor: Pungent Boiling Point: 90-91°/25mm Beilstein Reference: 1760574 UN No.: 3265 MDL No.: MFCD00039533

Formula: C₅H₇F₃O₂
Boiling Pt: 90…91 °C/25 mm
Storage Temperature: Ambient
MDL Number: MFCD00039533
CAS Number: 348-75-4
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR