(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol
Supplier: Thermo Scientific Chemicals
Synonyms:
1,2:5,6-Di-O-isopropylidene-D-mannitol, (1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol, (1S,2S)-1,2-BIS((R)-2,2-DIMETHYL-1,3-DIOXOLAN-4-YL)ETHANE-1,2-DIOL
L12127-06
L12127-14
CAAAAL12127-06EA
66.2
CAD
CAAAAL12127-06
CAAAAL12127-14
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol
CAS No.: 1707-77-3
Molecular Formula: C12H22O6
Formula Weight: 262.31
Storage Temperature: 2°C to 8°C
Melting Point: 120-122°
Beilstein Reference: 83765
Optical Rotation: +6° (c=5 in chloroform)
MDL No.: MFCD00003211
Formula:
C₁₂H₂₂O₆ MW: 262.302 g/mol Melting Pt: 120…122 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003211 CAS Number: 1707-77-3 EINECS: 216-954-8 |
Specification Test Results
Optical rotation | +6° (c=5 in chloroform) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...