Diethyl ethylidenemalonate 99%
Supplier: Thermo Scientific Chemicals
L12117-03
L12117-06
CAAAAL12117-03EA
42.98
CAD
CAAAAL12117-03
CAAAAL12117-06
Diethyl ethylidenemalonate 99%
Diethyl ethylidenemalonate
Fieser: 9,161
Purity: 99%
CAS No.: 1462-12-0
Molecular Formula: C9H14O4
Formula Weight: 186.21
Flash Point:
Formula:
C₉H₁₄O₄ Boiling Pt: 115…116 °C/17 mm Density: 1.019 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009145 CAS Number: 1462-12-0 EINECS: 215-965-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...