Thiophene-3-acetic acid hydrazide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Thien-3-yl)acetohydrazide
L12077-03
L12077-06
CAAAAL12077-03EA
126.72
CAD
CAAAAL12077-03
CAAAAL12077-06
Thiophene-3-acetic acid hydrazide 97%
Thiophene-3-acetic acid hydrazide
Purity: 97%
CAS No.: 175276-94-5
Molecular Formula: C6H8N2OS
Formula Weight: 156.21
Melting Point: 85-86°
Beilstein Reference: 120473
MDL No.: MFCD00052946
Formula:
C₆H₈N₂OS Melting Pt: 85…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052946 CAS Number: 175276-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...