N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12072-06
L12072-14
CAAAAL12072-06EA
112.97
CAD
CAAAAL12072-06
CAAAAL12072-14
N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate 97%
N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate
Purity: 97%
CAS No.: 82228-89-5
Molecular Formula: C13H13NO4S.xH2O
Formula Weight: 279.32(anhy)
Melting Point: 98-100°
Beilstein Reference: 5695963
MDL No.: MFCD08061351
Formula:
C₁₃H₁₃NO₄S·xH₂O Melting Pt: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD08061351 CAS Number: 82228-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...