1-(4-Methoxyphenyl)ethanol 95%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
4-Methoxy-alpha-methylbenzyl alcohol
L12062-14
L12062-22
CAAAAL12062-14EA
110.76
CAD
CAAAAL12062-14
CAAAAL12062-22
1-(4-Methoxyphenyl)ethanol 95%
1-(4-Methoxyphenyl)ethanol
Purity: 95%
CAS No.: 3319-15-1
Molecular Formula: C9H12O2
Formula Weight: 152.19
Flash Point: 74°(165°F)
Refractive Index: 1.5330
Boiling Point: 94-96°/1mm
Beilstein Reference: 2043521
MDL No.: MFCD00016857
Formula:
C₉H₁₂O₂ Boiling Pt: 94…96 °C/1 mm Density: 1.079 Flash Pt: 74 °C(165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00016857 CAS Number: 3319-15-1 EINECS: 222-019-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...