1H,1H,5H-Octafluoropentyl methacrylate 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning

L12044-06 L12044-14
CAAAAL12044-06EA 134.93 CAD
CAAAAL12044-06 CAAAAL12044-14
1H,1H,5H-Octafluoropentyl methacrylate 98% stabilized
1H,1H,5H-Octafluoropentyl methacrylate

Purity: 98% CAS No.: 355-93-1 Molecular Formula: C9H8F8O2 Formula Weight: 300.15 Flash Point: 81°(177°F) Refractive Index: 1.3600 Boiling Point: 178-179° Beilstein Reference: 1801409 MDL No.: MFCD00039278

Formula: C₉H₈F₈O₂
Boiling Pt: 178…179 °C
Density: 1.432
Flash Pt: 81 °C(177 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039278
CAS Number: 355-93-1
EINECS: 206-596-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR