4-Nitrophenyl octanoate 96%

Supplier: Thermo Scientific Chemicals

L12022-03 L12022-06
CAAAAL12022-03EA 46.29 CAD
CAAAAL12022-03 CAAAAL12022-06
4-Nitrophenyl octanoate 96%
4-Nitrophenyl octanoate

Purity: 97% CAS No.: 1956-10-1 Molecular Formula: C14H19NO4 Formula Weight: 265.31 Refractive Index: 1.5130 MDL No.: MFCD00024665

Formula: C₁₄H₁₉NO₄
Density: 1.082
Storage Temperature: Ambient
MDL Number: MFCD00024665
CAS Number: 1956-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR