1-Isocyanato-3-(methylthio)benzene 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(Methylthio)phenyl isocyanate

L12008-03 L12008-06
CAAAAL12008-03EA 75.21 CAD
CAAAAL12008-03 CAAAAL12008-06
1-Isocyanato-3-(methylthio)benzene 96%
1-Isocyanato-3-(methylthio)benzene

Purity: 96% CAS No.: 28479-19-8 Molecular Formula: C8H7NOS Formula Weight: 165.22 Storage Temperature: 2°C to 8°C Flash Point: 113°(235°F) Refractive Index: 1.6030 Boiling Point: 129-130°/5mm Beilstein Reference: 387507 UN No.: 2206 MDL No.: MFCD00013863

Formula: C₈H₇NOS
MW: 165.22 g/mol
Boiling Pt: 129…130 °C (5 mmHg)
Density: 1.199
Flash Pt: 113 °C (235 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00013863
CAS Number: 28479-19-8
UN: 2206
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR