Sodium-4-aminoazobenzene-4'-sulfonate 90%, tech.

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Aminoazobenzene-4'-sulfonic acid Sodium Salt, 4-Aminoazobenzene-4'-sulphonic acid Sodium Salt

L11965-14 L11965-22
CAAAAL11965-14EA 53.25 CAD
CAAAAL11965-14 CAAAAL11965-22
Sodium-4-aminoazobenzene-4'-sulfonate 90%, tech.
Sodium-4-aminoazobenzene-4'-sulfonate

Purity: 90% CAS No.: 2491-71-6 Molecular Formula: C12H10N3NaO3S Formula Weight: 299.29 Beilstein Reference: 8098548 MDL No.: MFCD00035564

Formula: C₁₂H₁₁N₃O₃S
Storage Temperature: Ambient
MDL Number: MFCD00035564
CAS Number: 2491-71-6
EINECS: 219-657-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR