n-Propyl bromoacetate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11951-06
L11951-14
CAAAAL11951-06EA
61.94
CAD
CAAAAL11951-06
CAAAAL11951-14
n-Propyl bromoacetate 97%
n-Propyl bromoacetate
Purity: 97%
CAS No.: 35223-80-4
Molecular Formula: BrCH2CO2(CH2)2CH3
Formula Weight: 181.04
Flash Point: 82°(179°F)
Refractive Index: 1.4510
Boiling Point: 175-176°
Beilstein Reference: 1745564
UN No.: 2922
MDL No.: MFCD00075247
Formula:
BrCH₂CO₂(CH₂)₂CH₃ Boiling Pt: 175…176 °C Density: 1.399 Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075247 CAS Number: 35223-80-4 UN: 2922 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...