Mono-n-dodecyl phosphate 90%, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11941-06
L11941-14
CAAAAL11941-06EA
175.38
CAD
CAAAAL11941-06
CAAAAL11941-14
Mono-n-dodecyl phosphate 90%, tech.
Mono-n-dodecyl phosphate
Purity: 90%
CAS No.: 2627-35-2
Molecular Formula: C12H27O4P
Formula Weight: 266.32
Odor: Odorless
Melting Point: 39-45°
Beilstein Reference: 1710518
MDL No.: MFCD00013946
Formula:
C₁₂H₂₇O₄P Melting Pt: 39…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013946 CAS Number: 2627-35-2 EINECS: 220-095-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...