2-Methoxyethyl acrylate 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 50 ppm-100 ppm
L11902-0B
L11902-18
L11902-30
CAAAAL11902-0BEA
225.94
CAD
CAAAAL11902-0B
CAAAAL11902-18
CAAAAL11902-30
2-Methoxyethyl acrylate 98% stabilized
2-Methoxyethyl acrylate
Stab. with ca 50-100ppm 4-methoxyphenol
Purity: 98%
CAS No.: 3121-61-7
Molecular Formula: C6H10O3
Formula Weight: 130.14
Flash Point: 56°(133°F)
Refractive Index: 1.4280
Odor: Ester-like
Melting Point:
Formula:
C₆H₁₀O₃ Boiling Pt: 162 °C Melting Pt: Density: 1.008 Flash Pt: 56 °C(133 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00048149 CAS Number: 3121-61-7 EINECS: 221-499-3 UN: 1992 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...