Myosmine, (max. 2% H₂O) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11886-MC
L11886-ME
CAAAAL11886-MCEA
129.4
CAD
CAAAAL11886-MC
CAAAAL11886-ME
Myosmine, (max. 2% H₂O) 98%
Myosmine
Purity: 98%
CAS No.: 532-12-7
Molecular Formula: C9H10N2
Formula Weight: 146.19
Melting Point: 42-44°
Boiling Point: 82-83°/0.5mm
Beilstein Reference: 119254
MDL No.: MFCD00052019
Formula:
C₉H₁₀N₂ Boiling Pt: 82…83 °C/0.5 mm Melting Pt: 42…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052019 CAS Number: 532-12-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...