N-(2-Hydroxypropyl)morpholine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Morpholino-2-propanol
L11859-09
L11859-18
CAAAAL11859-09EA
90.85
CAD
CAAAAL11859-09
CAAAAL11859-18
N-(2-Hydroxypropyl)morpholine 98%
N-(2-Hydroxypropyl)morpholine
Purity: 98%
CAS No.: 2109-66-2
Molecular Formula: C7H15NO2
Formula Weight: 145.20
Flash Point: 92°(197°F)
Refractive Index: 1.4635
Boiling Point: 218°
Beilstein Reference: 1443
MDL No.: MFCD00023378
Formula:
C₇H₁₅NO₂ Boiling Pt: 218 °C Density: 1.020 Flash Pt: 92 °C (197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00023378 CAS Number: 2109-66-2 EINECS: 218-298-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...