4',5'-Methylenedioxy-2'-nitroacetophenone 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-(6-Nitrobenzo[d][1,3]dioxol-5-yl)ethanone
L11841-03
L11841-06
CAAAAL11841-03EA
134.14
CAD
CAAAAL11841-03
CAAAAL11841-06
4',5'-Methylenedioxy-2'-nitroacetophenone 98%
4',5'-Methylenedioxy-2'-nitroacetophenone
Purity: 98%
CAS No.: 56136-84-6
Molecular Formula: C9H7NO5
Formula Weight: 209.16
Melting Point: 121-123°
Beilstein Reference: 199756
Formula:
C₉H₇NO₅ MW: 209.16 g/mol Melting Pt: 121…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053045 CAS Number: 56136-84-6 EINECS: 260-010-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...