Phenyl acrylate 97%

Supplier: Thermo Scientific Chemicals
Warning

L11819-06 L11819-14
CAAAAL11819-06EA 127.82 CAD
CAAAAL11819-06 CAAAAL11819-14
Phenyl acrylate 97%
Phenyl acrylate

Purity: 97% CAS No.: 937-41-7 Molecular Formula: C6H5O2CCH=CH2 Formula Weight: 148.16 Refractive Index: 1.5210 Boiling Point: 89-90°/12mm Beilstein Reference: 2041124 UN No.: 3082 MDL No.: MFCD00048145

Formula: C₉H₈O₂
Boiling Pt: 89…90 °C/12 mm
Density: 1.076
Storage Temperature: Ambient
MDL Number: MFCD00048145
CAS Number: 937-41-7
EINECS: 213-329-1
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR