5-Chloro-2,4-dinitrotoluene 97%

Supplier: Thermo Scientific Chemicals
Warning

L11808-06 L11808-14
CAAAAL11808-06EA 98.12 CAD
CAAAAL11808-06 CAAAAL11808-14
5-Chloro-2,4-dinitrotoluene 97%
5-Chloro-2,4-dinitrotoluene

Purity: 97% CAS No.: 51676-74-5 Molecular Formula: C7H5ClN2O4 Formula Weight: 216.58 Melting Point: 89-91° Beilstein Reference: 2506591 UN No.: 2811 MDL No.: MFCD00041235

Formula: C₇H₅ClN₂O₄
Melting Pt: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00041235
CAS Number: 51676-74-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR